Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Kcnj15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039056.2 | NP_001034145.1 | 4 | 521 | ||
NM_001039057.2 | NP_001034146.1 | 4 | 613 | |||
NM_001271687.1 | NP_001258616.1 | 5 | 685 | |||
NM_001271689.1 | NP_001258618.1 | 5 | 652 | |||
NM_001271690.1 | NP_001258619.1 | 5 | 613 | |||
NM_001271691.1 | NP_001258620.1 | 4 | 593 | |||
NM_001271692.1 | NP_001258621.1 | 4 | 581 | |||
NM_001271693.1 | NP_001258622.1 | 4 | 548 | |||
NM_001271694.1 | NP_001258623.1 | 4 | 536 | |||
NM_001271695.1 | NP_001258624.1 | 3 | 495 | |||
NM_019664.5 | NP_062638.1 | 5 | 697 | |||
GenBank mRNA | AJ012368.1 | - | 4 | 479 | ||
AJ300831.1 | - | 1 | 145 | |||
AK085689.1 | - | 3 | 355 | |||
AK143004.1 | - | 4 | 402 | |||
AK163000.1 | - | 3 | 571 | |||
AK165346.1 | - | 5 | 521 | |||
AK165436.1 | - | 4 | 522 | |||
AY377988.1 | - | 2 | 168 | |||
BC010794.1 | - | 4 | 387 | 4221516 | ||
BC057915.1 | - | 5 | 512 | 5042647 | ||
BC138477.1 | - | 3 | 394 | 8861497 | ||
BC138478.1 | - | 3 | 406 | 8861498 | ||
BC145288.1 | - | 3 | 394 | 9053842 | ||
CA491479.1 | - | 4 | 517 |