Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bloc1s6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019788.3 | NP_062762.1 | 5 | 582 | ||
GenBank mRNA | AK038381.1 | - | 5 | 560 | ||
AK077720.1 | - | 5 | 564 | |||
AK147116.1 | - | 5 | 585 | |||
AK160580.1 | - | 5 | 552 | |||
AK170964.1 | - | 5 | 513 | |||
AK172571.1 | - | 5 | 563 | |||
BC016554.1 | - | 5 | 561 | |||
BC085260.1 | - | 5 | 551 | 6437935 |