Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031190.2 | NP_112467.2 | 1 | 634 | ||
GenBank mRNA | AK015016.1 | - | 1 | 625 | ||
AK133436.1 | - | 1 | 623 | |||
BC052343.1 | - | 1 | 598 | 6773764 | ||
BC061054.1 | - | 1 | 625 | 6774063 | ||
M18654.1 | - | 1 | 580 |