Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146145.2 | NP_666257.2 | 5 | 690 | ||
GenBank mRNA | AK033004.1 | - | 1 | 1005 | ||
AK081552.1 | - | 5 | 665 | |||
AK133866.1 | - | 5 | 630 | |||
AK134143.1 | - | 5 | 633 | |||
AK141210.1 | - | 5 | 632 | |||
AK143650.1 | - | 5 | 635 | |||
AK148521.1 | - | 5 | 655 | |||
AK152128.1 | - | 5 | 653 | |||
AK180982.1 | - | 1 | 53 | |||
AK196334.1 | - | 1 | 53 | |||
BC039752.1 | - | 5 | 659 | |||
S63728.1 | - | 5 | 640 |