Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146145.2 | NP_666257.2 | 6, 7 | 971 | ||
GenBank mRNA | AK081552.1 | - | 6, 7 | 946 | ||
AK133866.1 | - | 6, 7 | 913 | |||
AK134143.1 | - | 6, 7 | 914 | |||
AK141210.1 | - | 6, 7 | 913 | |||
AK143650.1 | - | 6, 7 | 916 | |||
AK148521.1 | - | 6, 7 | 936 | |||
AK152128.1 | - | 6, 7 | 934 | |||
AK181063.1 | - | 1, 2 | 78 | |||
BC039752.1 | - | 6, 7 | 940 | |||
S63728.1 | - | 6, 7 | 921 |