Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201600.2 | NP_963894.1 | 3 | 461 | ||
GenBank mRNA | BC030481.1 | - | 3 | 472 | ||
BC118525.1 | - | 3 | 454 | 40099275 | ||
CJ052777.1 | - | 390 |