Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270362.1 | NP_001257291.1 | 2 | 221 | ||
NM_001270363.1 | NP_001257292.1 | 2 | 221 | |||
NM_005053.3 | NP_005044.1 | 2 | 221 | |||
NR_072976.1 | 2 | 221 | ||||
GenBank mRNA | AB209713.1 | - | 2 | 144 | ||
AK289908.1 | - | 2 | 158 | |||
AK293219.1 | - | 2 | 180 | |||
BC014026.2 | - | 2 | 147 | 4300551 | ||
BC088364.1 | - | 2 | 150 | 30389208 | ||
BI832600.1 | - | 2 | 195 | |||
BP213821.1 | - | 2 | 205 | |||
BX448989.2 | - | 2 | 143 | |||
D21235.1 | - | 2 | 122 | |||
DB456593.1 | - | 2 | 222 |