Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007929.2 | NP_031955.2 | 5 | 760 | ||
GenBank mRNA | AF083876.1 | - | 5 | 627 | ||
AF346627.1 | - | 5 | 702 | |||
AK076194.1 | - | 5 | 740 | |||
AK077260.1 | - | 5 | 763 | |||
BC031646.1 | - | 4 | 553 | |||
BC041670.1 | - | 5 | 663 | 3663158 |