Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007936.3 | NP_031962.2 | 8, 9 | 1774 | ||
GenBank mRNA | AK028509.1 | - | 8, 9 | 1767 | ||
AK147698.1 | - | 8, 9 | 1770 | |||
BC004782.1 | - | 8, 9 | 1689 | 3588285 | ||
BC052164.1 | - | 8, 9 | 1746 | 6512978 | ||
S57168.1 | - | 8, 9 | 1774 | |||
X65138.1 | - | 8, 9 | 1774 |