Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Etv2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007959.2 | NP_031985.2 | 1, 2 | 103 | ||
GenBank mRNA | AB221612.1 | - | 1, 2 | 86 | ||
L10427.1 | - | 1, 2 | 103 |