Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ewsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001283061.1 | NP_001269990.1 | 13 | 1388 | ||
NM_001283062.1 | NP_001269991.1 | 11 | 1274 | |||
NM_007968.3 | NP_031994.2 | 13 | 1385 | |||
GenBank mRNA | AK014366.1 | - | 5 | 354 | ||
AK019460.1 | - | 13 | 1334 | |||
AK034755.1 | - | 13 | 1361 | |||
AK049743.1 | - | 11 | 1254 | |||
AK147103.1 | - | 13 | 1352 | |||
AK150345.1 | - | 13 | 1346 | |||
AK151625.1 | - | 13 | 1356 | |||
AK153276.1 | - | 13 | 1346 | |||
AK200262.1 | - | 5 | 358 | |||
BC068226.1 | - | 13 | 1355 | 5707017 | ||
X79233.1 | - | 13 | 1374 |