Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007980.3 | NP_032006.1 | 3 | 600 | ||
GenBank mRNA | AK007995.1 | - | 3 | 298 | ||
AY523818.1 | - | 3 | 286 | |||
AY523819.1 | - | 3 | 286 | |||
BC013457.1 | - | 3 | 280 | 4218050 | ||
BY707992.1 | - | 3 | 296 | |||
M65034.1 | - | 3 | 294 |