Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007984.2 | NP_032010.2 | 1 | 204 | ||
GenBank mRNA | AK083935.1 | - | 1 | 204 | ||
AK167670.1 | - | 1 | 204 | |||
BC010338.1 | - | 1 | 190 | |||
BC029281.1 | - | 1 | 192 | |||
BC037137.1 | - | 1 | 201 | |||
BC052408.1 | - | 1 | 193 | 5716179 | ||
BY311024.1 | - | 1 | 206 | |||
L33726.1 | - | 1 | 188 |