Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007984.2 | NP_032010.2 | 5 | 1690 | ||
GenBank mRNA | AK083935.1 | - | 5 | 1691 | ||
AK167670.1 | - | 5 | 1690 | |||
BC052408.1 | - | 5 | 1679 | 5716179 | ||
L33726.1 | - | 5 | 1670 |