Gene Symbol: Fancc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042673.2 | NP_001036138.1 | 4 | 661 | ||
NM_001282942.1 | NP_001269871.1 | 3 | 464 | |||
NM_007985.3 | NP_032011.2 | 4 | 661 | |||
GenBank mRNA | AK035488.1 | - | 4 | 415 | ||
AK042226.1 | - | 4 | 631 | |||
AK080697.1 | - | 642 | ||||
AK082717.1 | - | 3 | 442 | |||
AK147025.1 | - | 4 | 641 | |||
AK163161.1 | - | 4 | 662 | |||
BC038609.1 | - | 3 | 446 | 5375091 | ||
L08266.1 | - | 4 | 642 |