- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Fancc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042673.2 | NP_001036138.1 | 14 | 1683 | ||
NM_001282942.1 | NP_001269871.1 | 13 | 1486 | |||
NM_007985.3 | NP_032011.2 | 14 | 1683 | |||
GenBank mRNA | AK035488.1 | - | 14 | 1437 | ||
AK042226.1 | - | 14 | 1653 | |||
AK082717.1 | - | 11 | 1204 | |||
AK147025.1 | - | 14 | 1663 | |||
BC038609.1 | - | 13 | 1468 | 5375091 | ||
L08266.1 | - | 14 | 1664 |