Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002777.1 | - | 5 | 754 | ||
AK010693.1 | - | 5 | 779 | |||
AK145400.1 | - | 5 | 772 | |||
AK145986.1 | - | 4 | 805 | |||
AK151047.1 | - | 3 | 566 | |||
AK154751.1 | - | 5 | 727 | |||
AK168333.1 | - | 5 | 763 | |||
AK169186.1 | - | 5 | 686 | |||
AK169242.1 | - | 5 | 770 | |||
BC004671.1 | - | 5 | 730 | 3498682 | ||
X60203.1 | - | 5 | 745 |