Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008027.2 | NP_032053.1 | 6 | 601 | ||
GenBank mRNA | AF145044.1 | - | 5 | 442 | ||
AK150692.1 | - | 6 | 623 | |||
AK150827.1 | - | 6 | 550 | |||
AK181092.1 | - | 3 | 271 | |||
AY167925.1 | - | 6 | 607 | |||
BC004647.1 | - | 6 | 611 | 3158071 | ||
U90435.1 | - | 6 | 601 |