Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008052.3 | NP_032078.2 | 10 | 3527 | ||
GenBank mRNA | AB015422.1 | - | 9 | 3130 | ||
AK088630.1 | - | 10 | 3483 | |||
AK169455.1 | - | 10 | 3528 | |||
AK220514.1 | - | 9 | 3108 | |||
BC053055.1 | - | 10 | 2673 | 5685525 | ||
BC065075.1 | - | 11 | 3309 | |||
U38252.1 | - | 8 | 2712 |