Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008057.3 | NP_032083.3 | 1 | 3051 | ||
GenBank mRNA | AK010239.2 | - | 1 | 632 | ||
BC049781.1 | - | 1 | 2774 | |||
BC063077.1 | - | 2 | 2382 | 6844727 |
Set Membership: |
![]() |