Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Gata3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008091.3 | NP_032117.1 | 6 | 2574 | ||
GenBank mRNA | AK090089.1 | - | 6 | 2585 | ||
AK157939.1 | - | 6 | 2438 | |||
BC062915.1 | - | 5 | 2171 | 6826352 |