Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077411.2 | NP_001070879.1 | 9 | 1385 | ||
NM_008094.5 | NP_032120.1 | 10 | 1554 | |||
GenBank mRNA | AK082767.1 | - | 9 | 1383 | ||
AK152745.1 | - | 9 | 1366 | |||
BC006663.1 | - | 10 | 1539 | 3498551 | ||
M24119.1 | - | 9 | 1294 |