Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ggt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079235.1 | - | 8 | 1035 | ||
BC012969.1 | - | 7 | 924 | 4037243 | ||
CT010196.1 | - | 6 | 782 | |||
U30509.1 | - | 7 | 942 |