Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008149.3 | NP_032175.2 | 5 | 484 | ||
GenBank mRNA | AK047295.1 | - | 5 | 485 | ||
AK137067.1 | - | 7 | 765 | |||
AK173202.1 | - | 5 | 509 | |||
BC019201.1 | - | 5 | 489 | 5041120 |