Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008187.2 | NP_032213.2 | 4 | 611 | ||
GenBank mRNA | AK088713.1 | - | 4 | 515 | ||
AK172603.1 | - | 4 | 587 | |||
BC092397.1 | - | 3 | 2216 | |||
BC119023.1 | - | 4 | 507 | 8733772 | ||
BC119025.1 | - | 4 | 507 | 8733774 | ||
BC144770.1 | - | 3 | 427 | |||
CB195345.1 | - | 1 | 33 | |||
Z54179.1 | - | 4 | 534 |