Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159410.1 | NP_001152882.1 | 8 | 890 | ||
NM_008193.3 | NP_032219.2 | 8 | 1053 | |||
GenBank mRNA | AK004205.1 | - | 8 | 912 | ||
AK013514.1 | - | 8 | 916 | |||
AV050819.1 | - | 1 | 241 | |||
BC024625.1 | - | 8 | 917 | 4485215 |