Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008214.4 | NP_032240.3 | 2 | 220 | ||
GenBank mRNA | AK011685.1 | - | 2 | 136 | ||
AK077437.1 | - | 2 | 136 | |||
AK088824.1 | - | 2 | 173 | |||
AK146922.1 | - | 2 | 173 | |||
AK150081.1 | - | 2 | 139 | |||
AK152329.1 | - | 2 | 152 | |||
AK166826.1 | - | 2 | 136 | |||
AK166892.1 | - | 2 | 183 | |||
AK211613.1 | - | 1 | 35 | |||
BC020088.1 | - | 2 | 155 | 4008479 | ||
CA561201.1 | - | 2 | 220 | |||
U39473.1 | - | 2 | 200 |