Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320866.1 | NP_001307795.1 | 7 | 891 | ||
NM_001721.6 | NP_001712.1 | 7 | 829 | |||
NM_203281.2 | NP_975010.1 | 7 | 891 | |||
XM_017029752.1 | XP_016885241.1 | 7 | 861 | |||
GenBank mRNA | AF045459.1 | - | 7 | 905 | ||
AK291824.1 | - | 7 | 861 | |||
AK313568.1 | - | 7 | 824 | |||
BC016652.2 | - | 7 | 846 | 3919253 | ||
X83107.1 | - | 7 | 751 |