Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hdgf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008231.4 | NP_032257.3 | 6 | 1746 | ||
GenBank mRNA | AF251787.1 | - | 6 | 1660 | ||
AK005359.1 | - | 6 | 1525 | |||
AK008393.1 | - | 2 | 752 | |||
AK008479.1 | - | 1 | 750 | |||
AK017863.1 | - | 6 | 1700 | |||
AK029475.1 | - | 6 | 1490 | |||
AK076021.1 | - | 6 | 1534 | |||
AK187445.1 | - | 1 | 285 | |||
BC005713.1 | - | 6 | 1657 | 2609261 | ||
BC021654.1 | - | 6 | 1675 | 4163897 | ||
D63707.1 | - | 6 | 1501 |