Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdgfrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK032572.1 | - | 2, 3 | 345 | ||
AK143616.1 | - | 2, 3 | 266 | |||
AK144669.1 | - | 2, 3 | 270 | |||
AK146813.1 | - | 2, 3 | 205 | |||
AK146918.1 | - | 2, 3 | 243 | |||
AK183373.1 | - | 1, 2 | 79 | |||
BC003741.1 | - | 2, 3 | 202 | 3488803 | ||
D63850.1 | - | 2, 3 | 246 | |||
FN687734.2 | - | 2, 3 | 148 |