Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160112.1 | NP_001153584.1 | 1 | 2612 | ||
NM_008241.2 | NP_032267.1 | 2 | 1574 | |||
GenBank mRNA | AK132305.1 | - | 1 | 2600 | ||
BC046958.1 | - | 1 | 1212 | 6314329 | ||
BC055774.1 | - | 1 | 906 | |||
BC064449.1 | - | 1 | 1517 | |||
BC079597.1 | - | 1 | 2278 | |||
U36760.1 | - | 2 | 1574 |