Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hoxb13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008267.3 | NP_032293.1 | 2 | 1198 | ||
GenBank mRNA | AK077273.1 | - | 2 | 1220 | ||
BC013639.1 | - | 2 | 1198 | 4207456 | ||
BC051087.1 | - | 1176 | 6530542 | |||
BC058813.1 | - | 2 | 1173 | 6530413 |