Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008308.4 | NP_032334.2 | 1 | 1897 | ||
GenBank mRNA | AK034017.1 | - | 1 | 1345 | ||
AK043668.1 | - | 1 | 1356 | |||
AK049814.1 | - | 1 | 1737 | |||
AK049884.1 | - | 1 | 1828 | |||
BC138669.1 | - | 1 | 1604 | 8861690 | ||
BC138681.1 | - | 1 | 1604 | 8861702 | ||
U39391.1 | - | 1 | 1405 |
Set Membership: |
![]() |