Search Thermo Fisher Scientific
Gene Symbol: Ifit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008332.3 | NP_032358.1 | 3 | 3682 | ||
GenBank mRNA | AK038027.1 | - | 3 | 3435 | ||
AK151472.1 | - | 1 | 1045 | |||
BC050835.1 | - | 3 | 3429 | 6513417 | ||
U43085.1 | - | 2 | 3257 |