Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010104.3 | NP_034234.1 | 3 | 948 | ||
GenBank mRNA | AB081657.1 | - | 3 | 334 | ||
AK040778.1 | - | 3 | 606 | |||
AK152647.1 | - | 3 | 599 | |||
AK153437.1 | - | 3 | 603 | |||
BC029547.1 | - | 3 | 742 | 5059522 | ||
D43775.1 | - | 3 | 659 | |||
U35233.1 | - | 3 | 353 |