Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010120.5 | NP_034250.3 | 3 | 1667 | ||
GenBank mRNA | AF026481.1 | - | 2 | 2488 | ||
AK009026.1 | - | 3 | 1638 | |||
AK013268.1 | - | 3 | 1666 | |||
AK019451.1 | - | 3 | 1670 | |||
AK150651.1 | - | 4 | 1593 | |||
AK150673.1 | - | 3 | 1640 | |||
AK150677.1 | - | 3 | 1640 | |||
AK166368.1 | - | 1639 | ||||
AK167333.1 | - | 3 | 1627 | |||
BC027437.1 | - | 3 | 1629 | 4017054 |