Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288627.1 | NP_001275556.1 | 5 | 628 | ||
NM_001288628.1 | NP_001275557.1 | 6 | 697 | |||
NM_010128.4 | NP_034258.1 | 5 | 586 | |||
GenBank mRNA | AK135453.1 | - | 3 | 301 | ||
AK157366.1 | - | 5 | 504 | |||
AK158369.1 | - | 5 | 525 | |||
AK159752.1 | - | 5 | 504 | |||
AK168806.1 | - | 5 | 478 | |||
AK171061.1 | - | 5 | 504 | |||
AK171675.1 | - | 5 | 504 | |||
AK172607.1 | - | 5 | 504 | |||
AK172705.1 | - | 5 | 504 | |||
BC034257.1 | - | 6 | 597 | 3157463 | ||
BC046299.1 | - | 5 | 586 | 6486958 | ||
BC050899.1 | - | 5 | 628 | 6402273 | ||
BC138491.1 | - | 5 | 447 | 8861511 | ||
CT010209.1 | - | 4 | 381 | |||
U25633.1 | - | 5 | 470 | |||
X98403.1 | - | 5 | 563 |