Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010130.4 | NP_034260.1 | 6 | 632 | ||
GenBank mRNA | AK133202.1 | - | 6 | 608 | ||
AK149898.1 | - | 6 | 618 | |||
AK151303.1 | - | 6 | 604 | |||
AK151682.1 | - | 6 | 624 | |||
AK155862.1 | - | 6 | 583 | |||
BC075688.1 | - | 6 | 604 | 30609706 | ||
U66888.1 | - | 6 | 619 | |||
U66889.1 | - | 6 | 619 | |||
U66891.1 | - | 4 | 343 | |||
U66892.1 | - | 3 | 196 | |||
X93328.1 | - | 6 | 628 |
Set Membership: |
![]() |