Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Epha2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010139.3 | NP_034269.2 | 17 | 3005 | ||
GenBank mRNA | AK137704.1 | - | 17 | 2987 | ||
AK144202.1 | - | 17 | 2989 | |||
BC140960.1 | - | 17 | 2961 | 9055572 | ||
X76010.1 | - | 10 | 1220 | |||
X78339.1 | - | 17 | 2967 |