Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fhl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010214.4 | NP_034344.2 | 2 | 2178 | ||
GenBank mRNA | AK005848.1 | - | 2 | 2149 | ||
AK029585.1 | - | 2181 | ||||
AK030256.1 | - | 2 | 2091 |