Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC23A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005116.5 | NP_005107.4 | 10 | 1287 | ||
NM_203327.1 | NP_976072.1 | 10 | 1305 | |||
XM_011529414.2 | XP_011527716.1 | 10 | 1292 | |||
XM_011529415.2 | XP_011527717.1 | 9 | 1113 | |||
XM_011529416.2 | XP_011527718.1 | 9 | 1422 | |||
XM_011529417.2 | XP_011527719.1 | 9 | 1169 | |||
XM_017028171.1 | XP_016883660.1 | 10 | 1326 | |||
XM_017028172.1 | XP_016883661.1 | 10 | 1330 | |||
XM_017028173.1 | XP_016883662.1 | 10 | 1310 | |||
XM_017028174.1 | XP_016883663.1 | 10 | 1205 | |||
XM_017028175.1 | XP_016883664.1 | 6 | 641 | |||
XM_017028176.1 | XP_016883665.1 | 7 | 959 | |||
XM_017028177.1 | XP_016883666.1 | 7 | 946 | |||
XM_017028178.1 | XP_016883667.1 | 9 | 1212 | |||
XM_017028179.1 | XP_016883668.1 | 9 | 1199 | |||
XM_017028180.1 | XP_016883669.1 | 5 | 523 | |||
XM_017028181.1 | XP_016883670.1 | 10 | 1304 | |||
GenBank mRNA | AF058319.1 | - | 8 | 912 | ||
AF092511.1 | - | 9 | 1208 | |||
AF164142.1 | - | 10 | 1305 | |||
AJ269478.1 | - | 8 | 912 | |||
AJ292318.1 | - | 9 | 1143 | |||
AK296304.1 | - | 7 | 953 | |||
AY380556.1 | - | 8 | 912 | |||
BC013112.2 | - | 10 | 1276 | 3923057 | ||
D87075.2 | - | 10 | 1287 | |||
EF032501.1 | - | 8 | 912 |