Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010477.4 | NP_034607.3 | 9 | 1179 | ||
GenBank mRNA | AK088844.1 | - | 9 | 1093 | ||
AK143882.1 | - | 9 | 1084 | |||
AK146831.1 | - | 9 | 1131 | |||
AK182202.1 | - | 194 | ||||
AK206160.1 | - | 3 | 193 | |||
AK208814.1 | - | 2 | 127 | |||
AK211782.1 | - | 3 | 193 | |||
AK212204.1 | - | 3 | 193 | |||
AK217294.1 | - | 3 | 193 | |||
BC016400.1 | - | 9 | 1065 | 3585270 | ||
BC018545.1 | - | 3 | 287 | |||
BC106112.1 | - | 9 | 1080 | |||
X55023.1 | - | 9 | 1041 |