Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2-M10.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013544.3 | NP_038572.3 | 3 | 531 | ||
GenBank mRNA | AF539597.1 | - | 3 | 531 | ||
AY211079.1 | - | 3 | 531 | |||
BC104320.1 | - | 3 | 397 | |||
BC104321.1 | - | 3 | 523 | |||
BC119458.2 | - | 3 | 531 | 40044798 | ||
BC152395.1 | - | 3 | 523 |