Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZRSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005089.3 | NP_005080.1 | 6, 7 | 479 | ||
XM_005274597.2 | XP_005274654.2 | 6, 7 | 447 | |||
XM_011545589.2 | XP_011543891.2 | 5, 6 | 411 | |||
XM_011545590.2 | XP_011543892.2 | 6, 7 | 1249 | |||
XM_017029881.1 | XP_016885370.1 | 6, 7 | 446 | |||
XM_017029882.1 | XP_016885371.1 | 5, 6 | 401 | |||
XM_017029883.1 | XP_016885372.1 | 6, 7 | 542 | |||
GenBank mRNA | AK309673.1 | - | 6, 7 | 446 | ||
BC050451.1 | - | 6, 7 | 450 | 6499970 | ||
BC113454.1 | - | 6, 7 | 479 | 8322506 | ||
BC113480.1 | - | 6, 7 | 479 | 8322532 | ||
D49677.1 | - | 6, 7 | 458 |