Search Thermo Fisher Scientific
Gene Symbol: Hp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017370.2 | NP_059066.1 | 5 | 748 | ||
GenBank mRNA | AK150430.1 | - | 5 | 641 | ||
AK150830.1 | - | 5 | 641 | |||
AK152295.1 | - | 5 | 641 | |||
AK152698.1 | - | 5 | 641 | |||
AK159676.1 | - | 5 | 640 | |||
BC138872.1 | - | 5 | 623 | 8861894 | ||
M96827.1 | - | 5 | 638 | |||
S67972.1 | - | 5 | 639 |