Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017371.2 | NP_059067.2 | 5 | 409 | ||
GenBank mRNA | AK145928.1 | - | 5 | 407 | ||
BC011246.1 | - | 5 | 389 | 4196684 | ||
BC019901.1 | - | 5 | 391 | 5123914 | ||
U89889.1 | - | 5 | 365 |