Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMYM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005095.2 | NP_005086.2 | 30 | 4548 | ||
XM_005271328.4 | XP_005271385.1 | 30 | 4827 | |||
XM_005271331.2 | XP_005271388.1 | 27 | 3793 | |||
XM_011542421.2 | XP_011540723.1 | 30 | 4474 | |||
XM_011542423.2 | XP_011540725.1 | 31 | 4879 | |||
XM_011542425.2 | XP_011540727.1 | 28 | 3833 | |||
XM_017002803.1 | XP_016858292.1 | 31 | 4601 | |||
XM_017002804.1 | XP_016858293.1 | 31 | 4541 | |||
XM_017002805.1 | XP_016858294.1 | 26 | 4039 | |||
GenBank mRNA | AB007885.1 | - | 26 | 3668 | ||
AK309705.1 | - | 29 | 4208 | |||
BC012093.1 | - | 28 | 4127 | |||
BC127113.1 | - | 31 | 4554 | 40125110 | ||
BC127114.1 | - | 27 | 3737 | 40125115 | ||
BX647376.1 | - | 26 | 3779 | |||
BX648067.1 | - | 21 | 2842 | |||
CR936741.1 | - | 29 | 4412 |