Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190984.1 | NP_001177913.1 | 6 | 754 | ||
NM_001190985.1 | NP_001177914.1 | 5 | 972 | |||
NM_021295.3 | NP_067270.1 | 6 | 788 | |||
GenBank mRNA | AJ294535.1 | - | 6 | 781 | ||
AK146120.1 | - | 5 | 963 | |||
BC038649.1 | - | 3 | 293 | |||
BC058560.1 | - | 6 | 754 | 6315377 | ||
Y11550.1 | - | 6 | 753 | |||
Y16518.1 | - | 5 | 932 |