Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Hpse | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152803.4 | NP_690016.1 | 4 | 576 | ||
GenBank mRNA | AF359507.1 | - | 4 | 565 | ||
AK040471.1 | - | 4 | 577 | |||
AK087283.1 | - | 4 | 603 | |||
AK154628.1 | - | 4 | 577 | |||
AY077467.1 | - | 4 | 568 | |||
AY151051.1 | - | 4 | 487 | |||
BC138782.1 | - | 4 | 570 | 8861804 | ||
BC138784.1 | - | 4 | 570 | 8861806 |