Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177301.5 | NP_796275.3 | 1 | 207 | ||
GenBank mRNA | AK085906.1 | - | 1 | 158 | ||
AK144684.1 | - | 1 | 161 | |||
BC027206.1 | - | 1 | 184 | 4218471 | ||
BC099683.1 | - | 1 | 202 | 6530863 |